DWKCT0010P
Azlon Hexagonal Base PMP Cylinders with Molded Graduations
class B, capacity 10 mL , with +/-0.2mL tolerance
Synonym(s):
CT0010P
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
feature
class B
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon CT0010P
capacity
10 mL , with +/-0.2mL tolerance
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for the measurement and dispensing of liquids in the laboratory
Features and Benefits
Conform to ISO 6706
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service