DWKBGE336P
Azlon Square Bottles
graduated, cap, Plug, translucent high-density polyethylene, capacity 500 mL
Synonym(s):
BGE336P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
translucent high-density polyethylene
feature
cap Plug
graduated
packaging
pkg of 10 ea
manufacturer/tradename
Azlon
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for storage of liquid and powder samples
Features and Benefits
Space saving square design
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service