Skip to Content
Merck
All Photos(2)

Key Documents

D188603

Sigma-Aldrich

2,5-Dimethylthiophene

98.5%

Synonym(s):

NSC 60689

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8S
CAS Number:
Molecular Weight:
112.19
Beilstein:
106450
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98.5%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

134 °C/740 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)s1

InChI

1S/C6H8S/c1-5-3-4-6(2)7-5/h3-4H,1-2H3

InChI key

GWQOOADXMVQEFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Restituto Tocmo et al.
Journal of agricultural and food chemistry, 62(23), 5296-5304 (2014-05-21)
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service