CAD9925B
Animal feeding needle, sterile, disposable, PTFE w/SS bite protector
size 15G, L × diam. 3.1 in. × 2.8 mm , ball
All Photos(2)
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
Recommended Products
size
15G
packaging
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
3.1 in. × 2.8 mm , ball
Related Categories
General description
- Flexible PTFE tube, covered at the hub end with type 304 stainless steel preventing tube damage due to biting during use
- PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
- Plastic Luer hubs and soft silicone tips
- Non-toxic and no-pyrogenic
- Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Choose from one of the most recent versions:
Certificates of Analysis (COA)