Key Documents
BAF420740002
SP Bel-Art® Secador® 4.0 Horizontal Profile Gas-Purge Desiccator Cabinet
clear, volume 1.9 ft3
About This Item
UNSPSC Code:
41104510
NACRES:
NB.77
material
clear
polycarbonate
feature
Can be stacked three high
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42074-0002
overall H × W × D
34.1 cm (13.4 in.) × 51.8 cm (20.4 in.) × 41.4 cm (16.3 in.)
volume
1.9 ft3
General description
This sturdy Secador® desiccator cabinet has a large, wide storage space and provides convenient, reliable dry storage of moisture and UV sensitive products. U.S. Patents: 6,834,920; 7,318,630 and D474,626.
  • Durastar co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack
  • Rear mounted gas ports are tapered to accept tubing with 1/4 to 3/8” I.D.
  • Easy-to-view digital hygrometer is located on the pull down “oven style” door; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Two removable, perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Stackable up to 3 units high with use of Stacking Brackets
  • 4.0 models can be converted from horizontal to vertical orientation by turning the unit and ordering vertical model shelves
  • Gas ports can be closed to use desiccator with standard desiccant cartridges
  • Overall Dimensions without gas ports: 51.8W x 41.4D x 34.1cmH (20.4 x 16.3 x 13.4")
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)