936189
AdQPhos
Synonym(s):
1,2,3,4,5-Pentaphenyl-1′-(di-adamantylphosphino)ferrocene
About This Item
Recommended Products
Assay
≥90%
Quality Level
reaction suitability
reaction type: Cross Couplings
functional group
phosphine
SMILES string
C=1C=CC(=CC1)C=23C4(C=5C=CC=CC5)=C6(C=7C=CC=CC7)[C-]8(C=9C=CC=CC9)C2(C=%10C=CC=CC%10)[Fe+2]%11%12%13%143468[CH]=%15[CH]%14=[CH]%13[C-]%12(P(C%16%17CC%18CC(CC(C%18)C%16)C%17)C%19%20CC%21CC(CC(C%21)C%19)C%20)[CH]%15%11
InChI
InChI=1S/C35H25.C25H34P.Fe/c1-6-16-26(17-7-1)31-32(27-18-8-2-9-19-27)34(29-22-12-4-13-23-29)35(30-24-14-5-15-25-30)33(31)28-20-10-3-11-21-28;1-2-4-23(3-1)26(24-11-17-5-18(12-24)7-19(6-17)13-24)25-14-20-8-21(15-25)10-22(9-20)16-25;/h1-25H;1-4,17-22H,5-16H2;/q2*-1;+2
InChI key
IQEGBICLOWIZDH-UHFFFAOYSA-N
General description
Further, by merging with micellar catalysis, this catalytic system demonstrated the feasibility of enabling α-arylation reactions under mild aqueous conditions. This approach provides an environmentally benign alternative to toxic/hazardous solvents like 1,4-dioxane or NMP, bringing modern organic synthesis closer to being ideally sustainable.
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service