935492
Potassium hexachlororuthenate(IV)
powder, 99.99% trace metals basis
Synonym(s):
Potassium hexachlororuthenate(IV), Potassium hexachlororuthenium, Potassium ruthenium chloride, Potassium ruthenium(IV) chloride
About This Item
Recommended Products
Quality Level
Assay
(Gravimetric analysis of Ru)
99.99% trace metals basis
form
powder
impurities
≤150 ppm trace metals
color
brown to very dark brown
pH
1-9
solubility
water: soluble
SMILES string
[K][K].Cl[Ru](Cl)(Cl)(Cl)(Cl)Cl
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service