Skip to Content
Merck
All Photos(3)

Key Documents

923990

Sigma-Aldrich

Bio-based Polyether Polyol

greener alternative

Mn 400-600 Da

Synonym(s):

α-Hydro-w-hydroxy, 1,3-Poly-propanediol, Bio-based PPD, PO3G Polypropanediol, Poly(oxy-1,3-propanediyl)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H6O)nH2O
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.22

mol wt

Mn 400-600 Da

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

concentration

≤500 ppm in water (QM-AA 1020-114)

viscosity

90-120 mPa.s(40 °C)

mp

0-5  °C

density

1.02 g/cm3 at 40 °C

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Bio-based polypropanediol (USDA 100% certified bio-based product) is low toxic, low volatile and bio-degradable. Compared to petrochemical alternatives such as poly-(tetramethylene-ether)glycol (PTMEG), polypropyleneglycol (PPG), polyethyleneglycol (PEG), polypropanediol has a significantly lower environmental footprint, saving 40 % in nonrenewable energy consumption and reducing greenhouse gas emissions by 42 % as proved by an ISO 14000-compliant life cycle analysis.
It is hydrolysis resistant, has a high oxidative stability and a high thermal capacity, which offers a long durability in end use applications. Among its potential applications:

  • High-performance elastomers - High-performace alternative to petroleum-based ingredients used in polyurethane , copolyamides and copolyesters elastomers. It allows an easy drop-in, soft-segment replacement for PTMEG thermoplastic elastomers, offering unique characteristics including good chemical resistance, high mechanical strength and toughness as well as increased softness and elastic recovery. Recently, a study showed that bio-based polypropanediol can be used for selective debonding of polyurethane-based adhesives at the end of their lifetime to facilitate the recycling of expensive components and raw materials from complex devices such as consumer electronics .
  • Coatings - It can be used as ingredient or additive for auto refinish, industrial metal coatings and PU dispersions in any or all of the coating layers, replacing any petroleum-based polyols. It provides excellent adhesion to metal, and offers outstanding color dispersion and chip resistance. When used as an additive in clear coating, it provides increased flexibility and improved gravel and flake resistance.
  • Ink-jet inks - When added to pigment-based ink-jet ink formulations, it combine the depth and clarity properties of dyes with the inherent durability of pigments.
  • Functional fluids - Thanks to its thermal conductivity, thermal stability and lubricity, excellent low temperature properties as well as low toxicity and inherent biodegradability, it is an ideal base fluid for lubricants, dielectric coolants and heat-transfer fluids.
  • Footwear and performance textile - Thanks to its superior abrasion resistance and Moisture Vapour Transmission Rate (MVTR), it offers applications in footwear and performance textile applications including synthetic leather.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to a family of high-performance polyether polyols based on renewable feedstock. Click here for more information.

Features and Benefits

Guideline:
  • Before usage, please keep the container sealed and stored at ambient temperature (5-25 °C).
  • For usage, it is recommended to work under nitrogen atmosphere.
  • In case it has to be molten, please do it at temperatures below 70 °C and under nitrogen atmosphere.
  • Conditions for safe storage, including any incompatibilities: Keep containers tightly closed in a cool, well-ventilated place. Store in original container and keep them away from heat and sources of ignition. Keep under nitrogen. Also keep away from food, drink and animal feedingstuffs.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Morales-Cerrada et al.
Polymers, 13(8) (2021-05-01)
The trend towards the utilization of bioresources for the manufacturing of polymers has led industry players to bring to the market new monomers. In this work, we studied 3 polyisocyanates and 2 polyols with high renewable carbon contents, namely L-lysine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service