Skip to Content
Merck
All Photos(1)

Key Documents

923052

Sigma-Aldrich

Copper(I) bromide

99.99% trace metals basis

Synonym(s):

Cuprous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.21

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: copper

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Copper (I) Bromide (CuBr) is a potential low cost, nontoxic hole transport layer material for organic solar cells. It is also a promising catalyst in combination with triethanolamine for atom transfer radical polymerization, especially in large scale applications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper Bromide as an Efficient Solution-Processable Hole Transport Layer for Organic Solar Cells: Effect of Solvents.
Bhargav R, et al.
ACS Omega, 4(3), 6028?6034-6028?6034 (2019)
CuBr/PMDETA combined with triethanolamine as an economic and highly active catalyst for atom transfer radical polymerization.
Zhang P, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 54(10), 735-741 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service