Skip to Content
Merck
All Photos(1)

Key Documents

911372

Lithium-7Li bromide

≥99 atom % 7Li, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
7LiBr
Molecular Weight:
86.92
UNSPSC Code:
41116105

isotopic purity

≥99 atom % 7Li

Assay

99% (CP)

form

solid

mass shift

depleted

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service