Skip to Content
Merck
All Photos(2)

Key Documents

910074

Sigma-Aldrich

27,27-Dimethyl-25-oxo-3,6,9,12,15,18,26-heptaoxaoctacosanoic acid

≥95%

Synonym(s):

2-2-2-2-2-2-6 PROTAC® linker, Bifunctional linker, OtBu-PEG6-acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44O10
Molecular Weight:
480.59
UNSPSC Code:
12352106

Assay

≥95%

form

liquid

reaction suitability

reagent type: linker

functional group

carboxylic acid
ester

storage temp.

2-8°C

SMILES string

O=C(CCCCCCOCCOCCOCCOCCOCCOCC(O)=O)OC(C)(C)C

Application

This heterobifunctional, PEGylated crosslinker 27,27-dimethyl-25-oxo-3,6,9,12,15,18,26-heptaoxaoctacosanoic acid features a carboxyl group at one end and a t-butyl protected carboxylic acid at the other and exhibits both hydrophobic and hydrophilic moieties for use in biological applications. The linker can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or pproteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation where the linker region is referred to as 2-2-2-2-2-2-6.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashton C Lai et al.
Angewandte Chemie (International ed. in English), 55(2), 807-810 (2015-11-26)
Proteolysis Targeting Chimera (PROTAC) technology is a rapidly emerging alternative therapeutic strategy with the potential to address many of the challenges currently faced in modern drug development programs. PROTAC technology employs small molecules that recruit target proteins for ubiquitination and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service