Skip to Content
Merck
All Photos(1)

Key Documents

87265

Sigma-Aldrich

Tetrafluorosuccinic anhydride

≥98.0% (T)

Synonym(s):

Perfluorosuccinic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4F4O3
CAS Number:
Molecular Weight:
172.03
Beilstein:
156388
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

liquid

refractive index

n20/D 1.327

bp

54-56 °C (lit.)

density

1.609 g/mL at 20 °C (lit.)

functional group

anhydride

SMILES string

FC1(F)C(=O)OC(=O)C1(F)F

InChI

1S/C4F4O3/c5-3(6)1(9)11-2(10)4(3,7)8

InChI key

ZLVLNNCBGQYRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service