807842
Copper
nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, ~5 mg/mL (in IPA)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
nanowires
Quality Level
concentration
~5 mg/mL (in IPA)
diam. × L
~100 nm (±20 nm) × 10~20 μm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Copper nanowires find applications in flexible electronics, transparent electrodes, photocatalysis, solar cells and as shape memory material [1, 2, 3].
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
53.6 °F
Flash Point(C)
12 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service