793892
15N2/O2/Ar Gas Mixture
ratio (10:20.95:69.05), 98 atom % 15N, 95% (CP)
About This Item
Recommended Products
isotopic purity
98 atom % 15N
Assay
95% (CP)
form
gas
composition
ratio (10:20.95:69.05)
1 of 4
This Item | 739731 | 431745 | 427691 |
---|---|---|---|
226-228 °C (lit.) | - | - | - |
n20/D 1.511 (lit.) | - | - | - |
1.138 g/mL at 25 °C (lit.) | - | - | - |
liquid | solid | solid | - |
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Gas 1 - Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service