76555
Pentadecanoyl chloride
≥98.0% (GC)
Synonym(s):
Pentadecanoic acid chloride
About This Item
Recommended Products
Assay
≥98.0% (GC)
form
liquid
refractive index
n20/D 1.451
density
0.909 g/mL at 20 °C
functional group
acyl chloride
SMILES string
CCCCCCCCCCCCCCC(Cl)=O
InChI
1S/C15H29ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3
InChI key
PQZWQGNQOVDTRF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service