Skip to Content
Merck
All Photos(1)

Key Documents

762989

Sigma-Aldrich

N,N-Diethyl-d10-formamide

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D10HNO
Molecular Weight:
111.21
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

liquid

mass shift

M+10

SMILES string

[H]C(N(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H])=O

InChI

1S/C5H11NO/c1-3-6(4-2)5-7/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2

InChI key

SUAKHGWARZSWIH-MWUKXHIBSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service