760609
Isocyanatophosphonic dichloride
95%
Synonym(s):
Dichloroisocyanatophosphine oxide, Dichlorophosphinyl isocyanate, Dichlorophosphonylisocyanate, Dichlorophosphoryl isocyanate, Isocyanatophosphonic acid dichloride, Isocyanatophosphoryl dichloride, Phosphine oxide, dichloroisocyanato-, Phosphoric dichloride isocyanate, Phosphorisocyanatidic dichloride, Phosphoroisocyanatidic dichloride, Phosphoryl dichloride isocyanate
About This Item
Recommended Products
Assay
95%
form
liquid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
refractive index
n20/D 1.468
density
1.623 g/mL at 25 °C
SMILES string
ClP(Cl)(=O)N=C=O
InChI
1S/CCl2NO2P/c2-7(3,6)4-1-5
InChI key
QKPIQPBNFUNZSJ-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
161.2 °F
Flash Point(C)
71.8 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service