754501
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline
Synonym(s):
(S)-4-(tert-butyl)-2-(pyridin-2-yl)-4,5-dihydrooxazole
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
Quality Level
mp
70-75 °C
SMILES string
CC(C)(C)[C@H]1COC(=N1)c2ccccn2
InChI
1S/C12H16N2O/c1-12(2,3)10-8-15-11(14-10)9-6-4-5-7-13-9/h4-7,10H,8H2,1-3H3/t10-/m1/s1
InChI key
JDOAIDZRVWJBEG-SNVBAGLBSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service