731382
(2R,2′R-(+)-[N,N′-Bis(2-pyridylmethyl)]-2,2′-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate
Synonym(s):
Fe(R,R-PDP), Iron(II) (R,R-(2-({(R)-2-[(R)-1-(pyridin-2-ylmethyl)pyrrolidin-2-yl]pyrrolidin-1-yl}methyl)pyridine)(bis-acetonitrile) hexafluoroantimonate, White-Chen catalyst, [Fe(R,R-PDP)(CH3CN)2](SbF6)2
About This Item
Recommended Products
form
powder
Quality Level
reaction suitability
core: iron
reagent type: catalyst
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
215-226 °C
greener alternative category
, Aligned
SMILES string
[Fe++].CC#N.CC#N.F[Sb-](F)(F)(F)(F)F.F[Sb-](F)(F)(F)(F)F.C1C[C@H]([C@H]2CCCN2Cc3ccccn3)N(C1)Cc4ccccn4
InChI
1S/C20H26N4.2C2H3N.12FH.Fe.2Sb/c1-3-11-21-17(7-1)15-23-13-5-9-19(23)20-10-6-14-24(20)16-18-8-2-4-12-22-18;2*1-2-3;;;;;;;;;;;;;;;/h1-4,7-8,11-12,19-20H,5-6,9-10,13-16H2;2*1H3;12*1H;;;/q;;;;;;;;;;;;;;;+2;2*+5/p-12/t19-,20-;;;;;;;;;;;;;;;;;/m1................./s1
InChI key
XUNWHBGEOCZDBX-QSKJABRXSA-B
General description
Application
Combined Effects on Selectivity in Fe-Catalyzed Methylene Oxidation
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service