Skip to Content
Merck
All Photos(1)

Key Documents

72962

Sigma-Aldrich

Methyl 4-nitrobenzoate

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CO2CH3
CAS Number:
Molecular Weight:
181.15
Beilstein:
1912198
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

flakes

mp

94-96 °C (lit.)
94-96 °C

functional group

ester

SMILES string

COC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H7NO4/c1-13-8(10)6-2-4-7(5-3-6)9(11)12/h2-5H,1H3

InChI key

YOJAHJGBFDPSDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service