Skip to Content
Merck
All Photos(1)

Key Documents

722243

Sigma-Aldrich

5-Methylbenzoxazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

40-47 °C

SMILES string

Cc1ccc2ocnc2c1

InChI

1S/C8H7NO/c1-6-2-3-8-7(4-6)9-5-10-8/h2-5H,1H3

InChI key

UBIAVBGIRDRQLD-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

190.0 °F - closed cup

Flash Point(C)

87.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen Larson et al.
The Journal of organic chemistry, 84(20), 13092-13103 (2019-09-25)
This manuscript details the development of the nickel-catalyzed arylation of oxazoles and benzoxazoles with aryl halides. A series of aryl, heteroaryl, and druglike electrophiles relevant to pharmaceutical applications were surveyed. The desired arylated products were obtained in synthetically useful yields

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service