Skip to Content
Merck
All Photos(1)

Key Documents

718408

Sigma-Aldrich

Trimethylsilyl cyanide-13C,15N

99 atom % 13C, 98 atom % 15N, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Si13C15N
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

97% (CP)

form

liquid

bp

114-117 °C (lit.)

mp

8-11 °C (lit.)

density

0.809 g/mL at 25 °C

mass shift

M+2

SMILES string

C[Si](C)(C)[13C]#[15N]

InChI

1S/C4H9NSi/c1-6(2,3)4-5/h1-3H3/i4+1,5+1

InChI key

LEIMLDGFXIOXMT-MQIHXRCWSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service