Skip to Content
Merck
All Photos(1)

Key Documents

705748

Sigma-Aldrich

4-Hydroxy-TEMPO-15N

98 atom % 15N, 97% (CP)

Synonym(s):

4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-15N-oxyl, TEMPOL-15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H1815NO2
CAS Number:
Molecular Weight:
173.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

97% (CP)

form

solid

mp

69-71 °C (lit.)

mass shift

M+1

storage temp.

2-8°C

SMILES string

CC1(C)CC(O)CC(C)(C)[15N]1[O]

InChI

1S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11H,5-6H2,1-4H3/i10+1

InChI key

UZFMOKQJFYMBGY-DETAZLGJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2 Oral

Target Organs

Liver,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service