Skip to Content
Merck
All Photos(1)

Key Documents

705403

Sigma-Aldrich

Nitrosobenzene-13C6

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5NO
CAS Number:
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

powder

mp

65-69 °C

mass shift

M+6

storage temp.

2-8°C

SMILES string

O=N[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C6H5NO/c8-7-6-4-2-1-3-5-6/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

NLRKCXQQSUWLCH-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service