Skip to Content
Merck
All Photos(1)

Key Documents

702595

Sigma-Aldrich

Zinc-70Zn gluconate

72 atom % 70Zn, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O1470Zn
Molecular Weight:
460.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

72 atom % 70Zn

Assay

97% (CP)

form

powder

mol wt

458.96 by atom % calculation (72 atom % 70Zn)

mp

172-175 °C (lit.)

mass shift

M+6

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[70Zn]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Zn/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;/h2*2-5,7-11H,1H2,(H,12,13);/q;;+2/p-2/t2*2-,3-,4+,5-;/m11./s1/i;;1+5

InChI key

WHMDKBIGKVEYHS-WKLCWQAVSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service