Skip to Content
Merck
All Photos(1)

Key Documents

680028

Sigma-Aldrich

Sitostanol-5,6,22,23-d4

97 atom % D, 95% (CP)

Synonym(s):

Stigmastan-5,6,22,23-d4-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29D4H48O
CAS Number:
Molecular Weight:
420.75
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

SMILES string

[2H]C1C[C@H]2[C@@H]3CC[C@H]([C@H](C)C([2H])C([2H])[C@@H](CC)C(C)C)[C@@]3(C)CC[C@@H]2[C@@]4(C)CC[C@H](O)C[C@]14[2H]

InChI

1S/C29H52O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h19-27,30H,7-18H2,1-6H3/t20-,21-,22+,23+,24+,25-,26+,27+,28+,29-/m1/s1/i8D,9D,10D,22D/t8?,9?,10?,20-,21-,22+,23+,24+,25-,26+,27+,28+,29-

InChI key

LGJMUZUPVCAVPU-DRBICYPTSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service