Skip to Content
Merck
All Photos(3)

Key Documents

666556

Sigma-Aldrich

6-Bromo-3-pyridinylboronic acid

≥95%

Synonym(s):

2-Bromopyridine-5-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(C5H3N)B(OH)2
CAS Number:
Molecular Weight:
201.81
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

161-166 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

OB(O)c1ccc(Br)nc1

InChI

1S/C5H5BBrNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3,9-10H

InChI key

BCYWDUVHAPHGIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride

Application

Reactant involved in:
  • Iterative cross-coupling reactions for synthesis of non-cytotoxic terpyridines
  • The Garlanding approach for synthesis of phenyl-pyridyl scaffolds
  • Cyanation for synthesis of aromatic nitriles
  • Suzuki-Miyaura cross-coupling with dihalopyridines and dihalobipyridines
  • Petasis boronic Mannich reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service