All Photos(1)
About This Item
Linear Formula:
HC≡CCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
96.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.407
density
0.728 g/mL at 25 °C
SMILES string
CC(C)CCC#C
InChI
1S/C7H12/c1-4-5-6-7(2)3/h1,7H,5-6H2,2-3H3
InChI key
HKNANEMUCJGPMS-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
19.4 °F
Flash Point(C)
-7 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service