633852
Ammonium-15N2 sulfate
15 atom % 15N
Synonym(s):
15N Labeled ammonium sulfate, Diazanium-15N2 sulfate
About This Item
Recommended Products
isotopic purity
15 atom % 15N
form
solid
mol wt
134.13 by atom % calculation (15 atom % 15N)
mp
>280 °C (dec.) (lit.)
application(s)
agriculture
mass shift
M+2
SMILES string
[15NH3].[15NH3].OS(O)(=O)=O
InChI
1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+1;
InChI key
BFNBIHQBYMNNAN-ISOLYIDJSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service