Skip to Content
Merck
All Photos(1)

Key Documents

611697

Sigma-Aldrich

Ethanol-d6

95% in D2O, 99 atom % D

Synonym(s):

Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2OD
CAS Number:
Molecular Weight:
52.11
Beilstein:
1699096
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

expl. lim.

3.5-15 % (lit.)

concentration

95% in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.358 (lit.)

bp

78 °C (lit.)

mp

-130 °C (lit.)

density

0.892 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2D2,3D

InChI key

LFQSCWFLJHTTHZ-LIDOUZCJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethanol-d6 may be used as a solvent for the tributylphosphine (PBu3) induced 5-exo cyclization of o-alkynoylphenol to form deuterated aurone.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethanol-induced changes in neuronal membrane order. An NMR study.
Hitzemann RJ, et al.
Biochimica et Biophysica Acta, 859(2), 189-197 (1986)
Johannes B Laurens et al.
Journal of forensic and legal medicine, 58, 155-163 (2018-07-08)
Sterile ante-mortem blood specimens were spiked with ethanol at the South African blood alcohol legal concentration limits of 0.20 g/L and 0.50 g/L and were stored in tubes containing sodium fluoride over a period of twenty-nine weeks under refrigeration (4 °C) and at
An Efficient Synthesis of Aurone Derivatives by the Tributylphosphine-catalyzed Regioselective Cyclization of o-Alkynoylphenols.
Saito K, et al.
Chemistry Letters (Jpn), 44(2), 141-143 (2015)

Protocols

GC/MS Analysis of Ethanol in Low Alcohol Beer on SUPELCOWAX® 10 after Headspace SPME using a 100 μm PDMS Fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service