605956
Taurine-13C2
≥99 atom % 13C, ≥98% (CP)
Synonym(s):
2-Aminoethane-13C2-sulfonic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2N13CH213CH2SO3H
CAS Number:
Molecular Weight:
127.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Recommended Products
isotopic purity
≥99 atom % 13C
Assay
≥98% (CP)
form
solid
technique(s)
bio NMR: suitable
mass spectrometry (MS): suitable
mp
>300 °C
mass shift
M+2
SMILES string
N[13CH2][13CH2]S(O)(=O)=O
InChI
1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)/i1+1,2+1
InChI key
XOAAWQZATWQOTB-ZDOIIHCHSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gil Sharon et al.
Cell, 177(6), 1600-1618 (2019-06-01)
Autism spectrum disorder (ASD) manifests as alterations in complex human behaviors including social communication and stereotypies. In addition to genetic risks, the gut microbiome differs between typically developing (TD) and ASD individuals, though it remains unclear whether the microbiome contributes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service