Skip to Content
Merck
All Photos(1)

Key Documents

604348

Sigma-Aldrich

2-Methyl-13C-furan

≥99 atom % 13C, ≥99% (CP), contains BHT as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC4H6O
CAS Number:
Molecular Weight:
83.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

BHT as stabilizer

bp

63-66 °C

density

0.921 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

[13CH3]c1ccco1

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3/i1+1

InChI key

VQKFNUFAXTZWDK-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Liver

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service