Skip to Content
Merck
All Photos(1)

Key Documents

577022

Sigma-Aldrich

Ethane-13C2/Helium Gas Mix

Ethane-13C2: Helium 1, Helium 9, 99% 13C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352001

isotopic purity

99% 13C

form

gas

composition

Ethane-13C2: Helium, 1
Helium, 9

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service