568155
4,4,5,5,-Tetramethyl-2-phenylsulfanylmethyl-1,3,2-dioxaborolane
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5SCH2BC2O2(CH3)4
CAS Number:
Molecular Weight:
250.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.5280 (lit.)
bp
105-108 °C/0.1 mmHg (lit.)
density
1.059 g/mL at 25 °C (lit.)
SMILES string
CC1(C)OB(CSc2ccccc2)OC1(C)C
InChI
1S/C13H19BO2S/c1-12(2)13(3,4)16-14(15-12)10-17-11-8-6-5-7-9-11/h5-9H,10H2,1-4H3
InChI key
DGPGLPBMZOKGON-UHFFFAOYSA-N
Related Categories
Application
4,4,5,5,-Tetramethyl-2-phenylsulfanylmethyl-1,3,2-dioxaborolane is used to prepare aryl/heteroaryl derivatives via the formation of C-C and C-hetero bonds.
It can also be used as:
It can also be used as:
- A starting material in the synthesis of α-aminoboronic acids as serine proteases inhibitors.
- A substrate in the coupling reactions with carbonyl compounds under Ir-catalyzed photoredox conditions.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of boronic acid analogues of α-amino acids by introducing side chains as electrophiles
Jagannathan S, et al.
The Journal of Organic Chemistry, 66(19), 6375-6380 (2001)
A Photoredox Coupling Reaction of Benzylboronic Esters and Carbonyl Compounds in Batch and Flow
Chen Y, et al.
Organic Letters, 21(15), 6140-6144 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service