All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.4710 (lit.)
bp
180-181 °C (lit.)
bulk density
0.896 g/mL
SMILES string
CC(C)N1CCNCC1
InChI
1S/C7H16N2/c1-7(2)9-5-3-8-4-6-9/h7-8H,3-6H2,1-2H3
InChI key
WHKWMTXTYKVFLK-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
129.9 °F - closed cup
Flash Point(C)
54.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service