558109
(R)-(−)-4-Phenyl-2-butanol
98%, optical purity ee: 99% (GLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5(CH2)2CH(OH)CH3
CAS Number:
Molecular Weight:
150.22
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
optical activity
[α]20/D −17.0°, c = 1% in chloroform
optical purity
ee: 99% (GLC)
refractive index
n20/D 1.5150 (lit.)
bp
206-207 °C (lit.)
density
0.976 g/mL at 25 °C (lit.)
functional group
hydroxyl
phenyl
SMILES string
C[C@@H](O)CCc1ccccc1
InChI
1S/C10H14O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3/t9-/m1/s1
InChI key
GDWRKZLROIFUML-SECBINFHSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service