Skip to Content
Merck
All Photos(1)

Key Documents

541451

Sigma-Aldrich

Poly(1-vinylnaphthalene)

average Mw ~30,000, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C10H7)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~30,000

transition temp

Tm 148 °C (DSC)

SMILES string

C=Cc1cccc2ccccc12

Looking for similar products? Visit Product Comparison Guide

Application

Used to measure optical Kerr effect. Also used as an efficient hole transporter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service