Skip to Content
Merck
All Photos(2)

Key Documents

535753

Sigma-Aldrich

2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol

powder

Synonym(s):

2-(2-Hydroxy-3,5-di-α-cumylphenyl)-2H -benzotriazole, 2-(2-Hydroxy-3,5-dicumyl)benzotriazole, 2-(2H -Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol, 2-(2H -Benzotriazol-2-yl)-4,6-bis(1-methyl-l-phenylethyl)phenol, 2-(3′,5′-Bis(α,α-dimethylbenzyl)-2′-hydroxyphenyl)benzotriazole, 2-(Benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol, 2-[2-Hydroxy-3,5-bis(α,α-dimethylbenzyl)phenyl]-2H -benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29N3O
CAS Number:
Molecular Weight:
447.57
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

loss

1.0 wt. % volatility, 264°C

mp

139-143 °C (lit.)

solubility

H2O: insoluble <0.04 ppm at 20 °C
toluene: 20 wt. % at 20 °C
methylene chloride: 34 wt. % at 20 °C
chloroform: 35 wt. % at 20 °C

SMILES string

CC(C)(c1ccccc1)c2cc(c(O)c(c2)C(C)(C)c3ccccc3)-n4nc5ccccc5n4

InChI

1S/C30H29N3O/c1-29(2,21-13-7-5-8-14-21)23-19-24(30(3,4)22-15-9-6-10-16-22)28(34)27(20-23)33-31-25-17-11-12-18-26(25)32-33/h5-20,34H,1-4H3

InChI key

OLFNXLXEGXRUOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Low volatile benzotriazole UV light absorber and stabilizer.
Imparts outstanding light stability for polymers processed at high temperatures such as polycarbonate, polyesters, polyacetal, polyamides, polyphenylene sulfide, polyphenylene oxide, aromatic copolymers, thermoplastic polyurethane and polyurethane fibers also highly effective for polyvinylchloride, styrene homo- and copolymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service