Skip to Content
Merck
All Photos(1)

Key Documents

49070

Sigma-Aldrich

(−)-Globulol

≥98.5% (sum of enantiomers, GC)

Synonym(s):

(1aR,4R,4aR,7R,7aS,7bS)-Decahydro-1,1,4,7-tetramethyl-1H-cycloprop[e]azulen-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
3195922
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D −45±2°, c = 1% in diethyl ether

mp

87-89 °C

functional group

hydroxyl

SMILES string

C[C@@H]1CC[C@@H]2[C@@H]1[C@H]3[C@@H](CC[C@@]2(C)O)C3(C)C

InChI

1S/C15H26O/c1-9-5-6-10-12(9)13-11(14(13,2)3)7-8-15(10,4)16/h9-13,16H,5-8H2,1-4H3/t9-,10-,11-,12-,13-,15-/m1/s1

InChI key

AYXPYQRXGNDJFU-QTPLKFIXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Globulol is a sesquiterpene that is isolated from Eucalyptus globulus Labill (Myrtaceae) fruits and leaves of Schinus areira. Globulol is a sweet, fruity, and green flavored chemical which can be found in different food products, including peppermint, orange mint, spearmint, and sweet bay. Globulol has also been reported as an active antioxidant and an antifungal agent.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Sesquiterpene Obtained from Schinus areira L.(Anacardiaceae) Essential Oil
Proceedings, 41(1), 85-85 (2019)
Antimicrobial activity of globulol isolated from the fruits of Eucalyptus globulus Labill
Tan M, et al.
Natural product research, 22(7), 569-575 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service