Skip to Content
Merck
All Photos(2)

Key Documents

423335

Sigma-Aldrich

2-(Methylthio)ethyl methacrylate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2CH2CH2SCH3
CAS Number:
Molecular Weight:
160.23
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

96%

refractive index

n20/D 1.48 (lit.)

bp

38 °C/0.06 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CSCCOC(=O)C(C)=C

InChI

1S/C7H12O2S/c1-6(2)7(8)9-4-5-10-3/h1,4-5H2,2-3H3

InChI key

MLGMNVSTCSILMV-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Conde-González et al.
Materials science & engineering. C, Materials for biological applications, 108, 110489-110489 (2020-01-12)
Three dimensional synthetic polymer scaffolds have remarkable chemical and mechanical tunability in addition to biocompatibility. However, the chemical and physical space is vast in view of the number of variables that can be altered e.g. chemical composition, porosity, pore size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service