Skip to Content
Merck
All Photos(1)

Key Documents

422975

Sigma-Aldrich

Ammonia-d3

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ND3
CAS Number:
Molecular Weight:
20.05
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor density

0.6 (vs air)

Quality Level

vapor pressure

4802 mmHg ( 15.5 °C)

isotopic purity

99 atom % D

Assay

99% (CP)

autoignition temp.

1203 °F

bp

−33 °C (lit.)

mp

−78 °C (lit.)

mass shift

M+3

SMILES string

[2H]N([2H])[2H]

InChI

1S/H3N/h1H3/i/hD3

InChI key

QGZKDVFQNNGYKY-ZRLBSURWSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard products are packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal pressure at 21ºC for both 10 L and 25 L standard package is 114 psig (liquified). This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Gas 2 - Press. Gas Liquefied gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service