Skip to Content
Merck
All Photos(2)

Key Documents

401684

Sigma-Aldrich

Lead(II) acetylacetonate

technical grade

Synonym(s):

2,4-Pentanedione lead(II) derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(C5H7O2)2
CAS Number:
Molecular Weight:
405.42
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

form

powder

reaction suitability

core: lead
reagent type: catalyst

mp

141-144 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[PbH2]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pb/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

UNNUWSQNTAFLDC-FDGPNNRMSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service