Skip to Content
Merck
All Photos(1)

Key Documents

285463

Sigma-Aldrich

D-Cystine

98%, for peptide synthesis

Synonym(s):

(S,S)-3,3′-Dithiobis(2-aminopropionic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728093
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

D-Cystine, 98%

Quality Level

Assay

98%

optical activity

[α]20/D +212°, c = 1 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

265 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](CSSC[C@@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m1/s1

InChI key

LEVWYRKDKASIDU-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Harada et al.
Journal of chromatography. A, 1593, 91-101 (2019-02-12)
Novel sophisticated derivatizing agents for the efficient enantioselective separation and mass spectrometric detection of d- and l-amino acids have been developed. Two new axially chiral reagents derived from 6,6'-dimethyl-2,2'-biphenyldiamine were synthesized. Their chiral separation and detection abilities were evaluated by
Theresa K Tiefenbrunn et al.
Protein science : a publication of the Protein Society, 18(5), 970-979 (2009-04-23)
The type 1 repeat domain from thrombospondin has potent antiangiogenic activity and a structurally interesting fold, making it an attractive target for protein engineering. Chemical synthesis is an attractive approach for studying protein domains because it enables the use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service