All Photos(3)
About This Item
Linear Formula:
(CH3)3CC6H4SO2Cl
CAS Number:
Molecular Weight:
232.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
78-81 °C (lit.)
SMILES string
CC(C)(C)c1ccc(cc1)S(Cl)(=O)=O
InChI
1S/C10H13ClO2S/c1-10(2,3)8-4-6-9(7-5-8)14(11,12)13/h4-7H,1-3H3
InChI key
YEZADZMMVHWFIY-UHFFFAOYSA-N
Related Categories
Application
4-tert-Butylbenzenesulfonyl chloride was used in the synthesis of 1-[bis-(4-fluorophenyl)-methyl]-4-(4-tert-butylbenzenesulfonyl)piperazine.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J N Narendra Sharath Chandra et al.
Bioorganic & medicinal chemistry, 14(19), 6621-6627 (2006-06-21)
A series of novel substituted 1-[bis(4-fluorophenyl)-methyl]piperazine derivatives (4a-g) and (5h-m) have been synthesized. The synthesized compounds were characterized by IR and 1H NMR. All the synthesized compounds were evaluated in vitro for their efficacy as antimicrobial agents against representative strains
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service