Skip to Content
Merck
All Photos(1)

Key Documents

246107

Sigma-Aldrich

Tribromofluoromethane

98%

Synonym(s):

Fluorotribromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CFBr3
CAS Number:
Molecular Weight:
270.72
Beilstein:
1732796
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.525 (lit.)

bp

106-107 °C (lit.)

mp

−73.6 °C (lit.)

density

2.765 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

FC(Br)(Br)Br

InChI

1S/CBr3F/c2-1(3,4)5

InChI key

IHZAEIHJPNTART-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tribromofluoromethane (fluorotribromomethane) was used in the synthesis of dibromofluoromethyllithium that reacted with an aldehyde or ketone to give fluorinated alcohol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation and carbonyl addition reactions of dibromofluoromethyllithium derived from tribromofluoromethane as applied to the stereoselective synthesis of fluoro olefins and 2-bromo-2-fluoro-1, 3-alkanediols.
Shimizu M, et al.
Bulletin of the Chemical Society of Japan, 71, 2903-2921 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service