Skip to Content
Merck
All Photos(1)

Key Documents

197475

Sigma-Aldrich

Ethyl dichlorophosphite

98%

Synonym(s):

Ethyl phosphorodichloridite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OPCl2
CAS Number:
Molecular Weight:
146.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

117-118 °C (lit.)

density

1.286 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(Cl)Cl

InChI

1S/C2H5Cl2OP/c1-2-5-6(3)4/h2H2,1H3

InChI key

PVCINRPAXRJLEP-UHFFFAOYSA-N

Application

Ethyl dichlorophosphite was used in the synthesis of peptide derivatives.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Use of Phosphorous Acid Chlorides in Peptide Synthesis1.
Young RW, et al.
Journal of the American Chemical Society, 78(10), 2126-2131 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service