185949
2-Chloro-2-methylpropane-d9
99 atom % D
Synonym(s):
tert-Butyl-d9 chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CD3)3CCl
CAS Number:
Molecular Weight:
101.62
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
vapor pressure
5.11 psi ( 20 °C)
isotopic purity
99 atom % D
Assay
99% (CP)
refractive index
n20/D 1.3819 (lit.)
bp
51-52 °C (lit.)
mp
-25 °C (lit.)
density
0.933 g/mL at 25 °C
mass shift
M+9
SMILES string
[2H]C([2H])([2H])C(Cl)(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C4H9Cl/c1-4(2,3)5/h1-3H3/i1D3,2D3,3D3
InChI key
NBRKLOOSMBRFMH-GQALSZNTSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Valerie L Colley et al.
Drug testing and analysis, 7(3), 241-246 (2014-10-11)
South American 'crack' cocaine, produced directly from coca leaf, can be distinguished from US domestically produced crack on the basis of occluded solvent profiles. In addition, analysis of domestically produced crack indicates the solvents that were used for cocaine hydrochloride
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service