Skip to Content
Merck
All Photos(1)

Key Documents

180912

Sigma-Aldrich

Poly(acrylonitrile-co-butadiene)

acrylonitrile 37-39 wt. %

Synonym(s):

1,3-Butadiene-acrylonitrile copolymer, Acrylonitrile-1,3-butadiene copolymer, Acrylonitrile-butadiene copolymer, Poly(acrylonitrile-butadiene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CN)]x(CH2CH=CHCH2)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

slab/chunk

Quality Level

composition

acrylonitrile, 37-39 wt. %

viscosity

70-80, Mooney(lit.)

density

1 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis, Characterization and Studies of Chemically Modified Poly (Acrylonitrile-Butadiene-Styrene) through Nitrile Functionalization: This paper explores the chemical modification of poly(acrylonitrile-butadiene-styrene) (ABS), particularly through nitrile functionalization to develop new materials with enhanced properties (George et al., 2023).
  • Poly (acrylonitrile-co-butadiene) as polymeric crosslinking accelerator for sulphur network formation: This study investigates the use of poly(acrylonitrile-co-butadiene) as a crosslinking accelerator in sulphur network formation, examining its impact on the mechanical properties of elastomers (Hait et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service