Skip to Content
Merck
All Photos(1)

Key Documents

175854

Sigma-Aldrich

Malonic acid-d4

98 atom % D, 99% (CP)

Synonym(s):

Propanedioic-2,2-d2 acid-1,3-d2, Tetradeuteriomalonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2(CO2D)2
CAS Number:
Molecular Weight:
108.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

132-135 °C (dec.) (lit.)

mass shift

M+4

SMILES string

[2H]OC(=O)C([2H])([2H])C(=O)O[2H]

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)/i1D2/hD2

InChI key

OFOBLEOULBTSOW-BGOGGDMHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service