135496
3-Amino-L-tyrosine dihydrochloride monohydrate
98%
Synonym(s):
3-Amino-L-tyrosine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H2NC6H3-4-(OH)CH2CH(NH2)CO2H · 2HCl · H2O
CAS Number:
Molecular Weight:
287.14
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
optical activity
[α]22/D −1.6°, c = 2 in H2O
reaction suitability
reaction type: solution phase peptide synthesis
mp
158-160 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
O.Cl.Cl.N[C@@H](Cc1ccc(O)c(N)c1)C(O)=O
InChI
1S/C9H12N2O3.2ClH.H2O/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14;;;/h1-3,7,12H,4,10-11H2,(H,13,14);2*1H;1H2/t7-;;;/m0.../s1
InChI key
ITQBVYBSXFBTJX-QTPLPEIMSA-N
Related Categories
General description
3-Amino-L-tyrosine dihydrochloride monohydrate also known as 3-Amino-L-tyrosine dihydrochloride, is commonly used in the solution-phase peptide synthesis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Katrin M Hinz et al.
Molecular endocrinology (Baltimore, Md.), 29(6), 933-942 (2015-05-07)
Thyroid hormones (THs) are transported across cell membranes by different transmembrane transporter proteins. In previous studies, we showed marked 3,3'-diiodothyronine (3,3'-T2) but moderate T3 uptake by the L-type amino acid transporter 2 (Lat2). We have now studied the structure-function relationships
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service